Skip to Content
Merck
All Photos(1)

Key Documents

T0328

Sigma-Aldrich

TAOK2 (1-314), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

KIAA0881, MAP3K17, PSK, PSK1, TAO1, TAO2

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$716.00

$716.00


Estimated to ship on25 April 2025


Request a Bulk Order

Select a Size

Change View
10 μG
$716.00

About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

$716.00


Estimated to ship on25 April 2025


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

88-120 nmol/min·mg

mol wt

~63 kDa

UniProt accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TAOK2(9344)

Biochem/physiol Actions

Thousand and one-amino acid protein kinase 2 (TAOK 2) is part of the stress-sensitive kinase cascade and activation of and binding to MKK3 by TAOK2 implicates it in the regulation of the p38-containing stress-responsive MAP kinase pathway. TAOK2 protein kinase activates p38 mitogen-activated protein kinase cascade in vitro and in cells by phosphorylating the MAP/ERK kinases MKK3 and MKK6. TAO2 may play an important role in the activation of specific intracellular signaling pathways that are unique to osmotic stress.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhu Chen et al.
The Journal of biological chemistry, 278(25), 22278-22283 (2003-04-01)
The TAO (for thousand-and-one amino acids) protein kinases activate p38 mitogen-activated protein (MAP) kinase cascades in vitro and in cells by phosphorylating the MAP/ERK kinases (MEKs) 3 and 6. We found that TAO2 activity was increased by carbachol and that
M Hutchison et al.
The Journal of biological chemistry, 273(44), 28625-28632 (1998-10-24)
Several components of the budding yeast pheromone-response pathway are conserved in mammalian mitogen-activated protein (MAP) kinase pathways. Thus, we used degenerate oligonucleotides derived from the sequence of the Saccharomyces cerevisiae protein kinase Ste20p to amplify related sequences from the rat.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service