SYNF43243L
Synthware™ three neck round bottom flask with angled side necks
bottle capacity 3000 mL, side joint: ST/NS 24/40
Synonym(s):
Synthware F43243L, 3-neck round flask
About This Item
Recommended Products
material
clear
Joint
side joint: ST/NS24/40
packaging
pack of 1 ea
manufacturer/tradename
Kemtech F43243L
bottle capacity
3000 mL
Related Categories
General description
Application
Features and Benefits
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service