Skip to Content
Merck
All Photos(1)

Key Documents

SYNB135118

Synthware Desiccator with lid and ceramic plate

bottle O.D. 180 mm, clear borosilicate glass 3.3

Synonym(s):

Synthware B135118, desiccator with Top Knob

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43
material:
clear borosilicate glass 3.3
Pricing and availability is not currently available.

material

clear borosilicate glass 3.3

packaging

pack of 1 ea

manufacturer/tradename

Kemtech B135118

bottle O.D.

180 mm

Looking for similar products? Visit Product Comparison Guide

General description

Desiccator, Clear, Id 180mm, Includes Lid & Ceramic Plate

Application

kemetch; synthware; desiccator

Features and Benefits

Organic Chemistry

Legal Information

Synthware is a trademark of Kemtech America Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service