Skip to Content
Merck
All Photos(1)

Key Documents

SRP8048

Sigma-Aldrich

PEDF mouse

recombinant, expressed in CHO cells, FLAG® tagged, >95% (SDS-PAGE)

Synonym(s):

Caspin, PC cell-derived growth factor, Pigment epithelium-derived factor, Stromal cell-derived factor 3

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$512.00

$512.00


Check Cart for Availability


Select a Size

Change View
10 μG
$512.00

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

$512.00


Check Cart for Availability

biological source

mouse

recombinant

expressed in CHO cells

tag

FLAG® tagged

Assay

>95% (SDS-PAGE)

form

lyophilized

mol wt

~47 kDa by SDS-PAGE

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

<0.01 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

Pigment epithelium-derived factor (PEDF) is a 47kDa secreted glycoprotein that belongs to the non-inhibitory serpin family group. PEDF is widely expressed in adult and fetal tissues, including brain, spinal cord, plasma, bone, prostate, pancreas, heart and lung. PEDF acts as an angiogenesis inhibitor with neurotrophic, immunomodulation and antitumor properties. It functions as anti-angiogenic agent by counterbalancing the proangiogenic effect of VEGF. PEDF is one of the most abundant proteins released by adipocytes and induces insulin resistance in adipocytes and human skeletal muscle cells. Recently, it has been reported that PEDF is sufficient to maintain the self-renewal of pluripotent human embryonic stem cells.

Physical form

Lyophilized from PBS.

Reconstitution

Reconstitute with 100 μL deionized water.

Other Notes

Mouse PEDF (aa 20-417) is fused at the C-terminus to a FLAG®-tag.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service