Skip to Content
Merck
All Photos(1)

Key Documents

SRP8038

Sigma-Aldrich

IL-38 mouse

>95% (SDS-PAGE), recombinant, expressed in E. coli, FLAG® tagged, lyophilized

Synonym(s):

CD223, Interleukin-1 family member 10, Interleukin-38

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$702.00

$702.00


Check Cart for Availability


Select a Size

Change View
10 μG
$702.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$702.00


Check Cart for Availability

Product Name

IL-38 mouse, recombinant, expressed in E. coli, FLAG® tagged, >95% (SDS-PAGE)

biological source

mouse

recombinant

expressed in E. coli

tag

FLAG® tagged

Assay

>95% (SDS-PAGE)

form

lyophilized

mol wt

~20 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

<0.01 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Il1f10(215274)

General description

IL-38 (IL-1F10) mRNA is expressed in heart, placenta, fetal liver, spleen, thymus and tonsil. The expression in a variety of immune tissues and similarity to IL-1Ra suggest a role of IL-1F10 in the inflammatory response.

Physical form

Lyophilized from PBS and 0.5% trehalose.

Reconstitution

Reconstitute with 100 μL deionized water.

Other Notes

IL-38 [IL-1F10] (mouse) (aa 3-152) is fused at the C-terminus to a FLAG-tag.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ina Rudloff et al.
Arthritis & rheumatology (Hoboken, N.J.), 67(12), 3219-3225 (2015-09-01)
Knowledge of interleukin-38 (IL-38), formerly IL-1 family member 10, is sparse, but Il1f10 polymorphisms are associated with inflammatory diseases, and recombinant IL-38 inhibits inflammatory responses similar to those reported in the context of systemic lupus erythematosus (SLE). We undertook this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service