Skip to Content
Merck
All Photos(1)

Key Documents

SRP6464

Sigma-Aldrich

ApoJ/Clusterin human

recombinant, expressed in HEK 293 cells, ≥92% (SDS-PAGE)

Synonym(s):

AAG4, Apolipoprotein J, CLI, CLU, KUB1, MGC24903, SGP-2, SGP2, SP-40, TRPM-2, TRPM2

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$553.00

$553.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$553.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$553.00


Check Cart for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥92% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 50.9 kDa
observed mol wt 34-37 kDa by SDS-PAGE

packaging

pkg of 10 μg
pkg of 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CLU(1191)

General description

Asp23-Glu449, α chain 24.5kDa +β chain 27_kDa.The recombinant rh CLUS / Clusterin (Asp23-Glu 449) was cleaved into α chain and β chain, which form a heterodimer linked by disulfide bonds.
DTT reduced. corresponding to the cleaved β chain, andβ chain respectively.
Native Apolipoprotein J (ApoJ), also named Clusterin, is a heavily glycosylated, 75-80 kDa disulfide-linked heterodimeric protein. Despite being cloned since 1989, no genuine function has been attributed to ApoJ so far. The protein has been reportedly implicated in several diverse physiological processes such as sperm maturation, lipid transportation, complement inhibition, tissue remodeling, and membrane recycling, cell-cell and cell-substratum interactions, stabilization of stressed proteins in a folding-competent state and promotion or inhibition of apoptosis. ApoJ gene is differentially regulated by cytokines, growth factors and stress-inducing agents. Clusterin is up- or down regulated on the mRNA or protein level in many pathological and clinically relevant situations including cancer, organ regeneration, infection, Alzheimer disease, retinitis pigmentosa, myocardial infarction, renal tubular damage, autoimmunity and others.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service