Skip to Content
Merck
All Photos(1)

Key Documents

SRP6420

Sigma-Aldrich

Kallikrein-1 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

KLK1, KLKR, Klk6, hK1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 27.9 kDa
observed mol wt 40-45 kDa (DTT-reduced. Protein migrates due to glycosylation. Pro 19 is the predicted N-terminal.)

packaging

pkg of 10 μg

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... KLK1(3816)

General description

Kallikrein-1 (KLK1) belongs to the peptidase S1 family and Kallikrein subfamily. KLK1 contains one peptidase S1 domain. KLK-1 preferential cleavage of Arg-|-Xaa bonds in small molecule substrates and also has highly selective action to release kallidin (lysyl-bradykinin) from kininogen involves hydrolysis of Met-|-Xaa or Leu-|-Xaa. Glandular Kallikreins cleave Met-Lys and Arg-Ser bonds in kininogen to release Lys-bradykinin.

Physical form

Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 2 mM CaCl2, 150 mM NaCl, pH 7.5. Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service