Skip to Content
Merck
All Photos(1)

Key Documents

SRP6399

Sigma-Aldrich

IL-2 R beta/CD122 human

recombinant, expressed in HEK 293 cells, ≥90% (SDS-PAGE)

Synonym(s):

IL2RB, P70-75, RP5-1170K4.6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥90% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 25.4 kDa
observed mol wt 40 kDa (DTT-reduced. Protein migrates due to glycosylation. Ala 27 is the predicted N-terminus.)

packaging

pkg of 10 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL2RB(3560)

General description

Interleukin-2 receptor (IL-2R)/ cluster of differentiation 122 (CD122) is a heterotrimeric protein, encoded by the gene mapped to human chromosome 22q12.3. The encoded protein is expressed on the surface of certain immune cells, such as lymphocytes, that binds and responds to a cytokine called IL-2. The IL-2R is made up of 3 subunits - α (CD25), β (CD122) and γ (CD132) - non-covalently associating. The α subunit is involved in binding IL-2, while signal transduction following cytokine interaction is carried out by the γ-chain, along with the β subunit. IL2R β is a member of the type I cytokine receptor family.

Application

Interleukin 2 receptor subunit β (IL2RB)/cluster of differentiation 122 (CD122) human has been used to study the effect of this cytokine on the expression of Klf15 in neurons.

Biochem/physiol Actions

Interleukin 2 receptor subunit β (IL2RB) is involved in signal transduction via its ligand interleukin-2. It is also implicated in inflammation. Cluster of differentiation 122 (CD122) is the receptor for interleukin-2 and is involved in receptor mediated endocytosis and transduces the mitogenic signals of IL2.

Physical form

Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cytokine receptor signaling through the Jak?Stat?Socs
pathway in disease.
Lynda A O, et al.
Molecular Immunology, 44, 2497-2506 (2007)
Autologous Regulatory T Cells for the Treatment of Type 1 Diabetes.
James A T, et al.
Current Diabetes Reports, 12(5), 623-632 (2012)
Abnormal expression pattern of the IL-2 receptor β-chain on CD4+ T cells in ANCA-associated vasculitis.
Wilde B, et al.
Disease Markers (2014)
Oxalate upregulates expression of IL-2Rβ and activates IL-2R signaling in HK-2 cells, a line of human renal epithelial cells.
Koul S, et al.
American Journal of Physiology: Renal Physiology, 306(9), F1039-F1046 (2014)
Edible Medicinal and Non Medicinal Plants: Volume 9, Modified Stems, Roots, Bulbs (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service