Skip to Content
Merck
All Photos(1)

Key Documents

SRP6362

Sigma-Aldrich

IL13RA2 human

recombinant, expressed in E. coli, ≥85% (SDS-PAGE)

Synonym(s):

CD213A2, CT19, IL-13R, IL13BP, Interleukin-13 receptor subunit alpha-2 precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥85% (SDS-PAGE)

form

liquid

mol wt

39.5 kDa (340 aa, 27-343 aa + His Tag)

packaging

pkg of 100 μg

concentration

1 mg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... IL13RA2(3598)

General description

IL13Rα2 (interleukin 13 receptor subunit alpha 2) is a 42 kDa monomeric high affinity IL13 receptor. This gene is located on human chromosome Xq.
IL13RA2 is closely related to Il13RA1, a subunit of the interleukin 13 receptor complex. This protein binds IL13 with high affinity, but lacks cytoplasmic domain, and does not appear to function as a signal mediator. It is reported to play a role in the internalization of IL13. Recombinant human IL13RA2 protein, fused to His-tag at N-terminus, was expressed in E.coli.

Biochem/physiol Actions

IL-13Rα2 (interleukin 13 receptor subunit α 2) induces the progression, migration and invasion of glioma cells. It has the ability to initiate Scr (selective catalytic reduction), phosphatidylinositol 3 kinase (PI3K), Akt (signaling pathway) and mTOR (mammalian target of rapamycin). IL-13Rα2 chain plays a key role in IL-13 cytotoxin-mediated therapy. It is expressed at high levels in several human tumors such as oral squamous cell carcinoma, breast cancer, clear cell renal cell carcinoma, hepatic cell cancer, ovarian cancer and colorectal cancer.

Physical form

1 mg/mL in 20 mM Tris-HCl buffer (pH 8.0) containing 0.4 M Urea and 10% glycerol.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interleukin-13 receptor α2 is associated with poor prognosis in patients with gastric cancer after gastrectomy.
Lin C, et al.
Oncotarget, 7(31), 49281-49281 (2016)
IL-13 receptor α2 stimulates human glioma cell growth and metastasis through the Src/PI3K/Akt/mTOR signaling pathway.
Tu M, et al.
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 37(11), 14701-14709 (2016)
Jinghan Fang et al.
Acta biomaterialia, 109, 95-108 (2020-04-09)
In order to elicit a desired barrier function in guided bone regeneration (GBR) or guided tissue regeneration (GTR), a barrier membrane has to maintain its integrity for a certain period of time to guarantee the regeneration of target tissue. Due
Polymorphisms in the IL13, IL13RA1, and IL4RA genes and rate of decline in lung function in smokers.
He JQ, et al.
American Journal of Respiratory Cell and Molecular Biology, 28(3), 379-385 (2003)
María Álvarez-Izquierdo et al.
Parasites & vectors, 13(1), 608-608 (2020-12-06)
The cytokine interleukin-25 (IL-25) is recognized as the most relevant initiator of protective T helper 2 (Th2) responses in intestinal helminth infections. This cytokine induces resistance against several species of intestinal helminths, including the trematode Echinostoma caproni. E. caproni has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service