Skip to Content
Merck
All Photos(1)

Documents

SRP6270

Sigma-Aldrich

MMP-2 human

recombinant, expressed in HEK 293 cells, ≥92% (SDS-PAGE)

Synonym(s):

CLG4, CLG4A, MMP-II, MMP2, MONA, TBE-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

tag

6-His tagged (C-terminus)

Assay

≥92% (SDS-PAGE)

form

lyophilized powder

mol wt

calculated mol wt 71.8 kDa
observed mol wt 66-71 kDa (DTT-reduced. Ala 30 is the predicted N-terminus.)

packaging

pkg of 10 and 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MMP-2(4313)

General description

Matrix metalloproteinase-2 (MMP-2) which is also known as 72kDa type IV collagenase, 72kDa gelatinase, gelatinase A and CLG4A, belongs to the peptidase M10A family. It contains three fibronectin type-II domains and four hemopexin-like domains. MMP-2 is produced by normal skin fibroblasts. The gene encoding this protein is localized on human chromosome 16q12.2.

Biochem/physiol Actions

Matrix metalloproteinase-2 (MMP-2) cleaves the collagen-like sequence Pro-Gln-Gly-|-Ile-Ala-Gly-Gln. It is involved in diverse functions such as remodeling of the vasculature, angiogenesis, tissue repair, tumor invasion, inflammation, and atherosclerotic plaque rupture. It degrades type IV, V, VII, and X collagens. The protein degrades extracellular matrix proteins and acts on several non-matrix proteins such as big endothelial 1 (bET) and β-type calcitonin gene-related peptide (CGRP) promoting vasoconstriction. MMP-2 also cleaves KISS (metastasis suppressor) at a Gly-|-Leu bond. The protein appears to have a role in myocardial cell death pathways and contributes to myocardial oxidative stress by regulating the activity of glycogen synthase kinase-3 β (GSK3β). It has been shown to cleave GSK3β in vitro. PEX, the C-terminal non-catalytic fragment of MMP2, possesses anti-angiogenic and anti-tumor properties and inhibits cell migration and cell adhesion to fibroblast growth factor-2 (FGF2) and vitronectin.

Physical form

Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 150 mM NaCl, pH 8.0. Generally 5-8% Mannitol or trehalose is added as a protectant before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matrix metalloproteinase-2 and myocardial oxidative stress injury: beyond the matrix.
Kandasamy AD
Cardiovascular Research, 85(3), 413-423 (2010)
Increased expression of 72-kd type IV collagenase (MMP-2) in human aortic atherosclerotic lesions.
Li Z
The American Journal of Pathology, 148(1), 121-128 (1996)
Disruption of angiogenesis by PEX, a noncatalytic metalloproteinase fragment with integrin binding activity.
Brooks PC,
Cell, 92(3), 391-400 (1998)
The association between inflammation-related genes and serum androgen levels in men: the prostate, lung, colorectal, and ovarian study.
Meyer TE
Prostate, 72(1), 65-71 (2012)
Activated interstitial myofibroblasts express catabolic enzymes and mediate matrix remodeling in myxomatous heart valves.
Rabkin E
Circulation, 104(21), 2525-2532 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service