Skip to Content
Merck
All Photos(1)

Key Documents

SRP6237

Sigma-Aldrich

SPARC/Osteonectin human

recombinant, expressed in CHO cells, ≥97% (SDS-PAGE)

Synonym(s):

BM-40, ON, Secreted protein acidic and rich in cysteine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

human

recombinant

expressed in CHO cells

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

mol wt

43.7 kDa

packaging

pkg of 10 μg
pkg of 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SPARC(6678)

General description

SPARC/Osteonectin is a secreted, evolutionarily conserved collagen-binding glycoprotein that is involved in a variety of cellular activities. It is highly expressed in tissues undergoing morphogenesis, remodeling and wound repair. SPARC/Osteonectin and its related peptides bind to numerous proteins of the extracellular matrix (ECM), affect ECM protein expression, influence cellular adhesion and migration, and modulate growth factor-induced cell proliferation and angiogenesis. SPARC/Osteonectin consists of three domains; an N-terminal acidic region that binds calcium ions with low affinity, a module containing two EF-hand motifs that bind calcium with high affinity, and a cysteine-rich follistatin-like domain. Recombinant human SPARC/Osteonectin is a glycoprotein containing 286 amino acids that migrates at an apparent MW of 43.7 kDa by SDSPAGE analysis due to the effect of glycosylation

Physical form

Sterile filtered through a 0.2 micron filter. Lyophilized from 10 mM Sodium Phosphate, pH 7.6.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service