Skip to Content
Merck
All Photos(1)

Key Documents

SRP6099

Sigma-Aldrich

GSTM1 from mouse

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

GST 1-1, GST class-mu1, Glutathione S-transferase Mu 1, pmGT10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

mouse

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

liquid

mol wt

25.9 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... GSTM1(14862)

General description

GSTM1 (glutathione S-transferase μ 1) belongs to the superfamily of glutathione-S-transferases. It belongs to the μ class of soluble GSTs. GSTM1 is strongly expressed in the kidneys.[1]

Biochem/physiol Actions

Members of GST (glutathione S-transferase) superfamily are responsible for the metabolism of various reactive oxygen species and xenobiotics. GSTM1 (GST μ 1) plays a significant role in vascular homeostasis. Humans with null allele, GSTM1(0), are at increased risks for colon and lung cancers, hypertension, and atherosclerosis, and have higher DNA changes in atherosclerotic lesions of the abdominal aorta.[1]

Physical form

1 mg/mL solution in PBS (pH 7.4) containing 5 mM Glutathione

Preparation Note

Centrifuge the vial prior to opening.

Other Notes

MPMILGYWNV RGLTHPIRML LEYTDSSYDE KRYTMGDAPD FDRSQWLNEK FKLGLDFPNL PYLIDGSHKI TQSNAILRYL ARKHHLDGET EEERIRADIV ENQVMDTRMQ LIMLCYNPDF EKQKPEFLKT IPEKMKLYSE FLGKRPWFAG DKVTYVDFLA YDILDQYRMF EPKCLDAFPN LRDFLARFEG LKKISAYMKS SRYIATPIFS KMAHWSNK

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glutathione S-transferase-micro1 regulates vascular smooth muscle cell proliferation, migration, and oxidative stress.
Yang Y, et al.
Hypertension, 54, 1360-1360 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service