Skip to Content
Merck
All Photos(1)

Key Documents

SRP6036

Sigma-Aldrich

CD-14 from mouse

recombinant, expressed in CHO cells, 90-95% (SDS-PAGE)

Synonym(s):

CD14, Cluster of differentiation 14

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

mouse

recombinant

expressed in CHO cells

Assay

90-95% (SDS-PAGE)

form

lyophilized

mol wt

50.0 kDa

packaging

pkg of 10 μg

impurities

Endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... CD14(12475)

General description

CD14 acts as a receptor for endotoxin (LPS), is expressed strongly on monocy¬tes, macrophages and weekly at surface of neutrophiles. CD14 is anchored to cells by linkage to glycosylphosphatidylinositol (GPI) and functions as a high affinity receptor LPS-LBP (lipopolysaccharide binding protein)-complexes. Furthermore CD14 is present in a soluble form in human serum, urine and other body fluids, acts at physiological concentrations as LPS agonist and has at higher concentrations an antagonizing effect in cell activation. The myeloid differentiation antigen CD14 acts as the major receptor for bacterial LPS. The dominant form of the recombinant wild type CD14 is the 50 kDa protein. This is a His-tagged protein.

Physical form

Lyophilized from a concentrated protein solution (1 mg/ml) containing PBS, pH 7.2

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in 10 μl of ultra-pure distilled water. This solution can then be diluted into other aqueous buffers.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service