Skip to Content
Merck
All Photos(1)

Key Documents

SRP5344

Sigma-Aldrich

CyclinK, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CCNK, CPR4, MGC9113

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$475.00

$475.00


Check Cart for Availability


Select a Size

Change View
20 μG
$475.00

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

$475.00


Check Cart for Availability

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~67 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CCNK(8812)

General description

CyclinK is a member of the cyclin family which can regulate transcription through their association with and activation of cyclin-dependent kinases (CDK). The CDK/CyclinK complex can phosphorylate the C-terminal domain (CTD) of the large subunit of RNA polymerase II. CyclinK is associated with potent CTD kinase and CDK kinase activities in vitro and coimmunoprecipitates with POLR2A. CyclinK is a new member of the transcription cyclin family which may play a dual role in regulating CDK and RNA polymerase II activities.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service