Skip to Content
Merck
All Photos(1)

Documents

SRP5340

Sigma-Aldrich

BCL2, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

Bcl-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~51 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... BCL2(596)

General description

BCL2 gene encodes an integral inner mitochondrial membrane protein that acts as an antiapoptotic protein. The protein BAD can antagonize both the cell cycle and antiapoptotic functions of BCL2 through binding to the BH3 domain. Constitutive expression of BCL2, such as in the case of translocation of BCL2 to Ig heavy chain locus, is thought to be the cause of follicular lymphoma. BCL2 is phosphorylated on specific serine/threonine residues within the unstructured loop in response to diverse stimuli and such phosphorylation has been associated with the loss of the biological function of BCL2.
It is a target gene of nuclear factor κB (NF-κB). The gene encoding this protein is localized on human chromosome 18q21. B-cell lymphoma 2 (BCL-2) inhibits apoptosis in cells and has been shown to be upregulated in adenomas.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Posttranslational modifications of Bcl2 family members--a potential therapeutic target for human malignancy.
Basu A
Frontiers in Bioscience, 11, 1508-1521 (2006)
BCL2 protein expression in follicular lymphomas with t(14;18) chromosomal translocations.
Masir N
British Journal of Haematology, 144(5), 716-725 (2009)
BCL2 family in DNA damage and cell cycle control.
Zingkel S
Cell Death and Differentiation, 13(8), 1351-1359 (2006)
P. Riederer, D.B. Calne
Advances in Research on Neurodegeneration null
Bcl-2 localized at the nuclear compartment induces apoptosis after transient overexpression.
Portier BP and Taglialatela G
The Journal of Biological Chemistry, 281(52), 40493-40502 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service