Skip to Content
Merck
All Photos(2)

Key Documents

SRP5305

Sigma-Aldrich

TNIK, active, GST tagged human

PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$508.00

$508.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
$508.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$508.00


Estimated to ship on30 May 2025


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO®

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

54-82 nmol/min·mg

mol wt

~67 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TNIK(23043)

General description

TNIK or TRAF2 and NCK interacting kinase is characterized by an N-terminal kinase domain and a C-terminal GCK domain that serves a regulatory function. TNIK is mainly expression in brain, heart, and spleen and it is a specific effector of RAP2 which regulate actin cytoskeleton. TNIK is autophosphorylated in a manner dependent upon lys54 in the ATP-binding pocket of its kinase domain and plays a main role in cytoskeleton regulation.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service