Skip to Content
Merck
All Photos(2)

Key Documents

SRP5301

Sigma-Aldrich

FGFR3 (K650E), active, GST tagged human

PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

ACH, CD333, CEK2, HSFGFR3EX, JTK4

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$508.00

$508.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$508.00

About This Item

UNSPSC Code:
51111800
NACRES:
NA.32

$508.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO®

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

11.2-16.8 nmol/min·mg

mol wt

~73 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... FGFR3(2261)

General description

Fibroblast growth factor receptor 3 (FGFR3) is part of a family of fibroblast growth factor receptors that share similar structure and function. FGFR3 plays a role in several important cellular processes, including regulation of cell growth and division, determination of cell fate, formation of blood vessels, wound healing and embryo development. FGFR3 is involved in the development and maintenance of bone and brain tissue. Mutations in FGFR3 have been implicated in causing bladder cancer, cancer of white blood cells (multiple myeloma) and cervical cancer.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service