Skip to Content
Merck
All Photos(2)

Key Documents

SRP5281

Sigma-Aldrich

SPHK1, active, GST tagged human

PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

SPHK

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$689.00

$689.00


Check Cart for Availability


Select a Size

Change View
10 μG
$689.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$689.00


Check Cart for Availability

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO®

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

465-592 nmol/min·mg

mol wt

~71 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SPHK1(8877)

General description

SPHK1 catalyzes the phosphorylation of sphingosine to form sphingosine-1-phosphate (SPP) which is a novel lipid messenger with both intracellular and extracellular functions. Various stimuli increase cellular levels of SPP by activation of SPHK1. Competitive inhibitors of SPHK1 block formation of SPP and selectively inhibit cellular proliferation induced by a variety of factors. SPHK1/sphingosine-1-phosphate pathway contributes to colon carcinogenesis. SPHK1 plays a key role in the generation and release of proinflammatory mediators triggered by anaphylatoxins from human macrophages and also is involved in the process of immune cell motility.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service