Skip to Content
Merck
All Photos(2)

Key Documents

SRP5274

Sigma-Aldrich

SIRT7, active, His tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MGC126840, MGC126842, SIR2L7

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$812.00

$812.00


Check Cart for Availability


Select a Size

Change View
10 μG
$812.00

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

$812.00


Check Cart for Availability

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

112-139 RLU/min·ng

mol wt

~46 kDa

NCBI accession no.

application(s)

genomic analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SIRT7(51547)

General description

SIRT7 is a member of the class IV of sirtuin family of proteins which are homologs to the yeast Sir2 protein and play a role in cell differentiation, proliferation, apoptosis, metabolism, and senescence. SIRT7 associates with active rRNA genes and histones. Overexpression of SIRT7 increases pol I-mediated transcription whereas knockdown of SIRT7 or inhibition of its catalytic activity results in decreased association of pol I with rDNA and reduced pol I transcription. Depletion of SIRT7 stops cell proliferation and triggers apoptosis. SIRT7 deacetylates p53 and increases cellular resistance to cytotoxic and oxidative stress.

Physical form

Supplied in 50 mM sodium phosphate, pH 7.0, 300 mM NaCl, 150 mM imidazole, 0.1 mM PMSF, 0.2 mM DTT, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valentina Sirri et al.
Journal of cell science, 132(17) (2019-07-25)
In humans, ribosome biogenesis mainly occurs in nucleoli following two alternative pre-rRNA processing pathways differing in the order in which cleavages take place but not by the sites of cleavage. To uncover the role of the nucleolar NAD+-dependent deacetylase sirtuin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service