Skip to Content
Merck
All Photos(2)

Key Documents

SRP5265

Sigma-Aldrich

HDAC1, active, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DKFZp686H12203, GON-10, HD1, RPD3, RPD3L1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$709.00

$709.00


Check Cart for Availability


Select a Size

Change View
10 μG
$709.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$709.00


Check Cart for Availability

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

7565-11765 RLU/min·ng

mol wt

~88 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HDAC1(3065)

General description

HDAC1 or Histone deacetylase 1 belongs to the histone deacetylase/acuc/apha family and is a component of the histone deacetylase complex which plays a key role in the regulation of eukaryotic gene expression. HDAC1 interacts with retinoblastoma tumor-suppressor protein and this complex is a key element in the control of cell proliferation and differentiation. Together with metastasis-associated protein-2, HDAC1 deacetylates p53 and modulates its effect on cell growth and apoptosis. HDAC1 is an essential element of the co-activation system for IFN-induced gene regulation and antiviral responses.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service