Skip to Content
Merck
All Photos(1)

Documents

SRP5260

Sigma-Aldrich

SETD8, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

PR-Set7, SET07, SET8

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~68 kDa

NCBI accession no.

application(s)

genomic analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SETD8(387893)

General description

SETD8 or SET domain containing lysine methyltransferase 8 is a lysine methyltransferase that monomethylates p53 in human cell lines. The monomethylation by SETD8 suppresses p53-mediated transcriptional activation of highly responsive target genes, such as p21 (CDKN1A) and PUMA (BBC3) but it has little influence on weak p53 targets. Depletion of SET8 augmented the proapoptotic and checkpoint activation functions of p53, and SETD8 expression is downregulated during DNA damage. SETD8 is a Wnt signaling mediator and is recruited by LEF1/TCF4 to regulate the transcription of Wnt-activated genes, possibly through H4K20 monomethylation at the target gene promoters.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service