Skip to Content
Merck
All Photos(2)

Key Documents

SRP5244

Sigma-Aldrich

MLK1 (1-433), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MAP3K9, MEKK9, PRKE1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$481.00

$481.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$481.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$481.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

99-133 nmol/min·mg

mol wt

~77 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MAP3K9(4293)

General description

MLK1 or Mixed-Lineage Kinase 1 is a mitogen-activated protein kinase kinase kinase capable of activating the c-Jun NH(2)-terminal kinase (JNK) pathway. The catalytic domain of MLK1 has amino acid sequence similarity to both the tyr-specific and the ser/thr-specific kinase classes. In addition, MLK1 contain 2 leu/ile-zipper motifs and a basic sequence near its C-termini. MLK1 is threonine (and possibly serine) phosphorylated at multiple sites in the activation loop, with phosphorylation of Thr312 required for full activation.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John T Durkin et al.
Biochemistry, 43(51), 16348-16355 (2004-12-22)
Mixed-lineage kinase 1 (MLK1) is a mitogen-activated protein kinase kinase kinase capable of activating the c-Jun NH(2)-terminal kinase (JNK) pathway. Full-length MLK1 has 1104 amino acids and a domain structure identical to MLK2 and MLK3. Immunoblot and mass spectrometry show
D S Dorow et al.
European journal of biochemistry, 213(2), 701-710 (1993-04-15)
Using the polymerase chain reaction to study mRNA expressed in human epithelial tumor cells, a member of a new family of protein kinases was identified. The catalytic domain of this kinase has amino-acid-sequence similarity to both the Tyr-specific and the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service