Skip to Content
Merck
All Photos(2)

Key Documents

SRP5233

Sigma-Aldrich

EPHA7 (580-end), active, GST tagged from mouse

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CEK11, EHK3, HEK11, MDK1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$481.00

$481.00


Estimated to ship on03 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
$481.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$481.00


Estimated to ship on03 May 2025


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

30-40 nmol/min·mg

mol wt

~73 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... Epha7(13841)

General description

EPHA7 is a member of the ephrin receptor subfamily of protein-tyrosine kinases family that have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2-fibronectin types III repeats. EPHA7 mediated signaling act as a physiologic trigger for apoptosis that can alter brain size and shape by regulating the number of neural progenitors. The high expression of EPHA7 protein plays an important role in the malignancy transformation, invasion progression and metastasis of primary hepatocellular carcinoma.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shui-jun Zhang et al.
Zhonghua wai ke za zhi [Chinese journal of surgery], 48(1), 53-56 (2010-03-23)
To investigate the expression and clinical significance of EphA7 protein in primary hepatocellular carcinoma. Immunohistochemistry and Western blot were used to detect the expression of EphA7 protein in 40 cases of primary hepatocellular carcinoma, their corresponding adjacent liver tissues and
Vanessa Depaepe et al.
Nature, 435(7046), 1244-1250 (2005-05-20)
Mechanisms controlling brain size include the regulation of neural progenitor cell proliferation, differentiation, survival and migration. Here we show that ephrin-A/EphA receptor signalling plays a key role in controlling the size of the mouse cerebral cortex by regulating cortical progenitor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service