Skip to Content
Merck
All Photos(2)

Key Documents

SRP5228

Sigma-Aldrich

PIM3 active, GST tagged from mouse

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

Kid1, MGC27707, MGC37517

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$545.00

$545.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
$545.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$545.00


Estimated to ship on29 May 2025


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

667-903 nmol/min·mg

mol wt

~63 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... Pim3(223775)

General description

PIM3 or pim-3 oncogene belongs to a family of protooncogenes that encode serine/threonine protein kinases. Pim-3 expressed in hepatocellular carcinoma development and plays a main role in the proliferation of human hepatoma cell lines. Pim-3 also expressed in human pancreatic cancer cell and phosphorylates BAD to block Bad-mediated apoptosis in human pancreatic cancer cell lines.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harald Mikkers et al.
Molecular and cellular biology, 24(13), 6104-6115 (2004-06-17)
The Pim family of proto-oncogenes encodes a distinct class of serine/threonine kinases consisting of PIM1, PIM2, and PIM3. Although the Pim genes are evolutionarily highly conserved, the contribution of PIM proteins to mammalian development is unclear. PIM1-deficient mice were previously
Ying-Yi Li et al.
Cancer research, 66(13), 6741-6747 (2006-07-05)
Pancreatic cancer still remains a serious health problem with <5% 5-year survival rate for all stages. To develop an effective treatment, it is necessary to identify a target molecule that is crucially involved in pancreatic tumor growth. We previously observed
Chifumi Fujii et al.
International journal of cancer, 114(2), 209-218 (2004-11-13)
Most cases of human hepatocellular carcinoma develop after persistent chronic infection with human hepatitis B virus or hepatitis C virus, and host responses are presumed to have major roles in this process. To recapitulate this process, we have developed the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service