Skip to Content
Merck
All Photos(1)

Key Documents

SRP5220

Sigma-Aldrich

KAT8 (2-467), GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

FLJ14040, KAT8, MOF, MYST1, hMOF

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$422.00

$422.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
20 μG
$422.00

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

$422.00


Check Cart for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~83 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... KAT8(84148)

General description

Lysine acetyltransferase 8 (KAT8) or MYST histone acetyltransferase 1 (MYST1) is a signaling protein that belongs to the MYST family of histone acetyl transferases (HATs). MYST1 has a chromodomain that is involved in protein-protein interactions and targeting transcriptional regulators to the chromatin.[1][2]

Biochem/physiol Actions

Lysine acetyltransferase 8 (KAT8) or MYST histone acetyltransferase 1 (MYST1) was originally isolated as a human immunodeficiency virus-1 (HIV-1) TAT-interactive protein, which plays important roles in regulating chromatin remodeling, transcription and other nuclear processes by acetylating histone and non-histone proteins. MYST1 is also involved in ataxia-telangiectasia mutated (ATM) function. It acetylates histone 4-lysine 16 (H4K16). The protein is a modulator of embryonic stem cells and an epigenetic regulator. It has been associated with gastric carcinoma.[3][1][4]

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Histone Acetyltransferase MOF Promotes Induces Generation of Pluripotent Stem Cells.
Mu X
Cellular Reprogramming, 17(4), 259-267 (2015)
MOF phosphorylation by ATM regulates 53BP1-mediated double-strand break repair pathway choice.
Gupta A
Cell Reports, 8(1), 177-189 (2014)
Expression of hMOF, but not HDAC4, is responsible for the global histone H4K16 acetylation in gastric carcinoma.
Zhu L
International Journal of Oncology, 46(6), 2535-2545 (2015)
Arun Gupta et al.
Molecular and cellular biology, 25(12), 5292-5305 (2005-06-01)
We have determined that hMOF, the human ortholog of the Drosophila MOF gene (males absent on the first), encoding a protein with histone acetyltransferase activity, interacts with the ATM (ataxia-telangiectasia-mutated) protein. Cellular exposure to ionizing radiation (IR) enhances hMOF-dependent acetylation
K C Neal et al.
Biochimica et biophysica acta, 1490(1-2), 170-174 (2000-04-29)
We have identified a novel human gene product, hMOF, which exhibits significant similarity to the Drosophila dosage compensation regulator, MOF. A recombinant C-terminal portion of hMOF has histone acetyltransferase activity directed toward histones H3, H2A and H4, a specificity characteristic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service