Skip to Content
Merck
All Photos(2)

Key Documents

SRP5217

Sigma-Aldrich

VRK1, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MGC117401, MGC138280, MGC142070

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$641.00

$641.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$641.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$641.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

1.7-2.3 nmol/min·mg

mol wt

~71 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... VRK1(7443)

General description

VRK1 is a member of the vaccinia-related kinase (VRK) family of serine/threonine protein kinases. VRK1 is widely expressed in human tissues and actively dividing cells, such as those in testis, leukocytes, fetal liver and carcinomas. VRK1 regulate cell proliferation and phosphorylates histone, casein, and the transcription factors ATF2 (activating transcription factor 2) and c-JUN. The spinal muscular atrophy with pontocerebellar hypoplasia is caused by a mutation in the VRK1 gene.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Nezu et al.
Genomics, 45(2), 327-331 (1997-11-05)
A cDNA library enriched for human fetal-specific liver genes was constructed by suppressive subtractive hybridization. EST fls223 generated from this library was found to represent a novel putative serine/threonine (Ser/Thr) kinase. A full-length clone isolated for this gene encodes a
Paul Renbaum et al.
American journal of human genetics, 85(2), 281-289 (2009-08-04)
The spinal muscular atrophies (SMAs) are a genetically and clinically heterogeneous group of disorders characterized by degeneration and loss of anterior horn cells in the spinal cord, leading to muscle weakness and atrophy. Spinal muscular atrophy with pontocerebellar hypoplasia (SMA-PCH

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service