Skip to Content
Merck
All Photos(1)

Key Documents

SRP5144

Sigma-Aldrich

Stathmin, His tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

C1orf215, FLJ32206, LAP18, Lag, MGC138869, MGC138870, OP18, PP17, PP19, PR22, SMN, STMN1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$426.00

$426.00


Estimated to ship on29 April 2025


Request a Bulk Order

Select a Size

Change View
50 μG
$426.00

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

$426.00


Estimated to ship on29 April 2025


Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~19 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... STMN1(3925)

Physical form

Supplied in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

STMN1 overexpression correlates with biological behavior in human cutaneous squamous cell carcinoma.
Li X
Pathology Research and Practice, 211(11), 816-823 (2015)
Covalent modification of stathmin by CCNU determined by FTMS analysis of modified proteins and tryptic peptides.
Wu WW
Biochemical and Biophysical Research Communications, 367(1), 7-13 (2008)
The stathmin phosphoprotein family: intracellular localization and effects on the microtubule network.
Gavet O
Journal of Cell Science, 111(Pt 22), 3333-3346 (1998)
Microtubule depolymerization contributes to spontaneous neurotransmitter release in vitro.
Velasco, et al.
Communications biology, 6, 488-488 (2023)
A Sobel
Trends in biochemical sciences, 16(8), 301-305 (1991-08-01)
Stathmin is a ubiquitous, phylogenetically conserved protein present in the cytoplasm of cells in a variety of unphosphorylated and phosphorylated forms. Its expression and phosphorylation are regulated throughout development and in response to extracellular signals regulating cell proliferation, differentiation and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service