Skip to Content
Merck
All Photos(1)

Key Documents

SRP5132

Sigma-Aldrich

SMAD3, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DKFZP586N0721, DKFZp686J10186, HSPC193, HsT17436, JV15-2, MADH3, MGC60396

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$364.00

$364.00


Estimated to ship on27 April 2025


Request a Bulk Order

Select a Size

Change View
50 μG
$364.00

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

$364.00


Estimated to ship on27 April 2025


Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~77 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SMAD3(4088)

General description

SMAD3 is a direct mediator of transcriptional activation by the TGFβ receptor. The activity of SMAD3 is regulated by the TGFβ receptors, and SMAD3 is phosphorylated and associated with the ligand-bound receptor complex. TGFβ stimulation leads to phosphorylation and activation of SMAD3, which form a complex with SMAD4 that accumulate in the nucleus and regulate transcription of target genes such as CDK inhibitor. SMAD3 containing a C-terminal truncation acts as a dominant-negative inhibitor of the normal TGFβ response. SMAD3 is a major physiologic substrate of the G1 cyclin-dependent kinases CDK4 and CDK2.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gareth J Inman et al.
Molecular cell, 10(2), 283-294 (2002-08-23)
Transforming growth factor (TGF)-beta stimulation leads to phosphorylation and activation of Smad2 and Smad3, which form complexes with Smad4 that accumulate in the nucleus and regulate transcription of target genes. Here we demonstrate that, following TGF-beta stimulation of epithelial cells
Isao Matsuura et al.
Nature, 430(6996), 226-231 (2004-07-09)
Transforming growth factor-beta (TGF-beta) potently inhibits cell cycle progression at the G1 phase. Smad3 has a key function in mediating the TGF-beta growth-inhibitory response. Here we show that Smad3 is a major physiological substrate of the G1 cyclin-dependent kinases CDK4

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service