Skip to Content
Merck
All Photos(2)

Key Documents

SRP5078

Sigma-Aldrich

PTPRA (174-802), active, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

HEPTP, HLPR, HPTPα, HPTPA, PTPA, PTPRL2, R-PTP-α, RPTPA

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$481.00

$481.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$481.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$481.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

1799-2435 nmol/min·mg

mol wt

~96 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PTPRA(5786)

General description

PTPRA (also known as LRP) is a member of the protein tyrosine phosphatase family and contains an extracellular domain, a single transmembrane segment and two tandem intracytoplasmic catalytic domains. PTPRA dephosphorylates SRC family kinases, potassium channels and NMDA receptors and exists as three alternatively spliced variants which encode two distinct isoforms. Absence of PTPRA compromises correct positioning of pyramidal neurons during development of mouse hippocampus.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angiola Petrone et al.
The EMBO journal, 22(16), 4121-4131 (2003-08-13)
Despite clear indications of their importance in lower organisms, the contributions of protein tyrosine phosphatases (PTPs) to development or function of the mammalian nervous system have been poorly explored. In vitro studies have indicated that receptor protein tyrosine phosphatase alpha
R J Matthews et al.
Proceedings of the National Academy of Sciences of the United States of America, 87(12), 4444-4448 (1990-06-01)
Protein-tyrosine-phosphatases (protein-tyrosine-phosphate phosphohydrolase, EC 3.13.48) have been implicated in the regulation of cell growth; however, to date few tyrosine phosphatases have been characterized. To identify additional family members, the cDNA for the human tyrosine phosphatase leukocyte common antigen (LCA; CD45)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service