Skip to Content
Merck
All Photos(2)

Key Documents

SRP5068

Sigma-Aldrich

PKN2/PRK2, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MGC150606, MGC71074, PAK2, PKN2, PRK2, PRKCL2, PRO2042, Pak-2

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$545.00

$545.00


Estimated to ship on27 April 2025


Request a Bulk Order

Select a Size

Change View
10 μG
$545.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$545.00


Estimated to ship on27 April 2025


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

63-85 nmol/min·mg

mol wt

~145 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PKN2(5586)

General description

PKN2 (also known as PRK2) is a Rho effector and a member of the protein kinase C superfamily of serine/threonine kinases. PKN2 is an essential regulator of both entry into mitosis and exit from cytokinesis in HeLa S3 cells. PKN2 is required for abscission of the midbody at the end of the cell division cycle and for phosphorylation and activation of Cdc25B, the phosphatase required for activation of mitotic cyclin/Cdk1 complexes at the G2/M transition. C-terminus of PKN2 could be a potential drug target for effector-specific pharmacological intervention of Rho-medicated biological processes.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anja Schmidt et al.
The EMBO journal, 26(6), 1624-1636 (2007-03-03)
Rho GTPases regulate multiple signal transduction pathways that influence many aspects of cell behaviour, including migration, morphology, polarity and cell cycle. Through their ability to control the assembly and organization of the actin and microtubule cytoskeletons, Rho and Cdc42 make
Wee Guan Lim et al.
Archives of biochemistry and biophysics, 479(2), 170-178 (2008-10-07)
PRK2/PKNgamma is a Rho effector and a member of the protein kinase C superfamily of serine/threonine kinases. Here, we explore the structure-function relationship between various motifs in the C-terminal half of PRK2 and its kinase activity and regulation. We report

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service