Skip to Content
Merck
All Photos(2)

Key Documents

SRP5064

Sigma-Aldrich

PI4K2B, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

FLJ11105, PI4KIIB, PIK42B

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$545.00

$545.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
10 μG
$545.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$545.00


Estimated to ship on01 June 2025


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

185-251 nmol/min·mg

mol wt

~84 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PI4K2B(55300)

General description

PI4K2B is a member of the phosphatidylinositol 4-kinase family (PI4K) that phosphorylate phosphatidylinositol to generate phosphatidylinositol 4-phosphate which are immediate precursor of several important signaling and scaffolding molecules. PI4K2B is primarily a cytosolic PI4K that is recruited to membranes where it stimulates phosphatidylinositol 4,5-bisphosphate syntheses. PI4K2B uses phosphatidylinositol as their primary substrate and has no activity on phosphatidylinositol monophosphates. PI4K2B is a restricted minor histocompatibility antigen in patients that had been successfully treated with donor lymphocyte infusions for relapsed chronic myeloid leukemia after allogeneic stem cell transplantation.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marieke Griffioen et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(10), 3837-3842 (2008-03-05)
Patients with hematological malignancies can be successfully treated with HLA-matched T cell-depleted allogeneic stem cell transplantation (alloSCT) and subsequent donor lymphocyte infusions (DLIs). The efficacy of DLI is mediated by donor T cells recognizing minor histocompatibility antigens (mHags) on malignant
Andras Balla et al.
The Journal of biological chemistry, 277(22), 20041-20050 (2002-03-30)
Phosphorylation of phosphatidylinositol (PI) to PI 4-phosphate is one of the key reactions in the production of phosphoinositides, lipid regulators of several cellular functions. This reaction is catalyzed by multiple enzymes that belong either to the type II or the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service