Skip to Content
Merck
All Photos(2)

Key Documents

SRP5052

Sigma-Aldrich

NEK3, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

HSPK36, MGC29949

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$481.00

$481.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$481.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$481.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

112-152 nmol/min·mg

mol wt

~86 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NEK3(4752)

General description

NEK3 is a member of the NEK family of protein kinases that share high amino acid homology with NIMA (never in mitosis gene a). NEK3 mRNA is detected in all the proliferating cell lines with the amount not changing during the cell cycle. During Prolactin receptor signaling, VAV2 is phosphorylated and activated by NEK3. Overexpression of NEK3 in Chinese hamster ovary cells increases cytoskeletal re-organization in response to Prolactin while downregulation of NEK3 expression by siRNA block these effects. Prolactin also stimulates interaction between NEK3 and paxillin leading to increased paxillin phosphorylation, Analysis of breast tissue microarrays show a significant up-regulation of NEK3 expression in malignant versus normal specimens.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S L Miller et al.
Oncogene, 26(32), 4668-4678 (2007-02-14)
Prolactin (PRL) stimulates the cytoskeletal re-organization and motility of breast cancer cells. During PRL receptor signaling, Vav2 becomes phosphorylated and activated, an event regulated by the serine/threonine kinase Nek3. Given the regulatory role of Vav2, the function of Nek3 in
M Kimura et al.
Cytogenetics and cell genetics, 95(3-4), 177-182 (2002-06-14)
NEKs (NIMA-related kinases) are a group of protein kinases sharing high amino acid sequence identities with NIMA (never in mitosis gene a) which control mitosis in Aspergillus nidulans. We have cloned a cDNA for human NEK3, a novel human gene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service