Skip to Content
Merck
All Photos(2)

Key Documents

SRP5047

Sigma-Aldrich

MEKK1 (664-1512), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MAP3K1, MAPKKK1, MEKK

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$641.00

$641.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
$641.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$641.00


Estimated to ship on24 May 2025


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

697-943 nmol/min·mg

mol wt

~155 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... Map3K1(4214)

General description

MEKK1 or MEK kinase, is a serine/threonine kinase that is downstream of mitogenic and metabolic stimuli, including insulin and many growth factors. MEKK1 functions not only as an upstream activator of ERK and JNK through its kinase domain, but also as an E3 ligase through its PHD domain, providing a negative regulatory mechanism for decreasing ERK1/ERK2 activity by ubiquitination and degradation. MEKK1 -/- embryonic stem cells from mice show loss or altered responses of JNK to microtubule disruption and cold stress. Furthermore, activation of JNK is lost and that of ERK is diminished in response to hyperosmolarity and serum factors in MEKK1 -/- cells.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Yujiri et al.
Science (New York, N.Y.), 282(5395), 1911-1914 (1998-12-04)
Targeted disruption of the gene encoding MEK kinase 1 (MEKK1), a mitogen-activated protein kinase (MAPK) kinase kinase, defined its function in the regulation of MAPK pathways and cell survival. MEKK1(-/-) embryonic stem cells from mice had lost or altered responses
Zhimin Lu et al.
Molecular cell, 9(5), 945-956 (2002-06-07)
ERK1/2 MAP kinases are important regulators in cellular signaling, whose activity is normally reversibly regulated by threonine-tyrosine phosphorylation. In contrast, we have found that stress-induced ERK1/2 activity is downregulated by ubiquitin/proteasome-mediated degradation of ERK1/2. The PHD domain of MEKK1, a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service