Skip to Content
Merck
All Photos(2)

Key Documents

SRP5045

Sigma-Aldrich

MARK2, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

EMK1, MGC99619, PAR-1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$481.00

$481.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$481.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$481.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

676-914 nmol/min·mg

mol wt

~114 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MARK2(2011)

General description

MARK2 or microtubule affinity-regulating kinase 2 is a member of the Par-1 family of serine/threonine protein kinases. MARK2 is an important regulator of cell polarity in epithelial and neuronal cells and controls the stability of microtubules through phosphorylation and inactivation of several microtubule-associating proteins. MARK2 is a key target of H. pylori CagA in the disorganization of gastric epithelial architecture underlying mucosal damage, inflammation, and carcinogenesis. MARK2 phosphorylates kinesin-like motor protein GAKIN/KIF13B to regulate axon formation. MARK2 function in the establishment of T cell polarity following engagement to an APC.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iraj Saadat et al.
Nature, 447(7142), 330-333 (2007-05-18)
Helicobacter pylori cagA-positive strains are associated with gastritis, ulcerations and gastric adenocarcinoma. CagA is delivered into gastric epithelial cells and, on tyrosine phosphorylation, specifically binds and activates the SHP2 oncoprotein, thereby inducing the formation of an elongated cell shape known
Takeshi Terabayashi et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 27(48), 13098-13107 (2007-11-30)
Neurons are highly polarized cells that possess two morphologically and functionally different types of protrusions, axons and dendrites, that function in the transmission and reception of neural signals, respectively. A great deal of attention has been paid to the specification
Zarin Tabassum et al.
The Journal of biological chemistry, 298(6), 101977-101977 (2022-04-27)
The posttranslational regulation of the neuronal proteome is critical for brain homeostasis but becomes dysregulated in the aged or diseased brain, in which abnormal posttranslational modifications (PTMs) are frequently observed. While the full extent of modified substrates that comprise the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service