Skip to Content
Merck
All Photos(2)

Key Documents

SRP5034

Sigma-Aldrich

GRK2, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

β-ARK1, ADRBK1, BARK1, FLJ16718

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$641.00

$641.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
$641.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

$641.00


Estimated to ship on18 May 2025


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

4.5-6.1 nmol/min·mg

mol wt

~102 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ADRBK1(156)

General description

G-protein-coupled receptor kinase 2 (GRK2),[1] which is officially known as adrenergic, beta, receptor kinase 1 (ADRBK1),[2] gene codes for a 689 amino acid protein, that is of 79.573kDa. This gene has 12 catalytic region subdomains. GRK2 is expressed in the vascular endothelium, arterial smooth muscle, myocardium, kidney and in the renal proximal tubule. It is mapped to human chromosome 11q13.[2]

Biochem/physiol Actions

G-protein-coupled receptor kinase 2 (GRK2) or ADRBK1 is a ubiquitous cytosolic enzyme that specifically phosphorylates the activated form of the beta-adrenergic and related G protein-coupled receptors. GRK2 phosphorylates the beta-2-adrenergic receptor and appears to mediate agonist-specific desensitization. Abnormal coupling of beta-adrenergic receptor to G protein is involved in the pathogenesis of the failing heart. RAF kinase inhibitor protein RKIP is a physiologic inhibitor of GRK2. After stimulation of G protein-coupled receptors, RKIP dissociates from its known target, RAF1 to associate with GRK2 and block its activity. G-protein-coupled receptor kinase 2 (GRK2) controls several intracellular mechanisms. It helps to maintain the structure and activities of the heart. Hence it participates in several cardiovascular diseases.[1] GRK2 also modulates ion and fluid transport and eventually, blood pressure. It increases the synthesis of mitochondrial superoxide and reduces oxygen utilization and ATP synthesis.[2] This protein serves as a major onco-modulator.[3]

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H A Rockman et al.
Proceedings of the National Academy of Sciences of the United States of America, 95(12), 7000-7005 (1998-06-17)
Heart failure is accompanied by severely impaired beta-adrenergic receptor (betaAR) function, which includes loss of betaAR density and functional uncoupling of remaining receptors. An important mechanism for the rapid desensitization of betaAR function is agonist-stimulated receptor phosphorylation by the betaAR
Kristina Lorenz et al.
Nature, 426(6966), 574-579 (2003-12-05)
Feedback inhibition is a fundamental principle in signal transduction allowing rapid adaptation to different stimuli. In mammalian cells, the major feedback inhibitor for G-protein-coupled receptors (GPCR) is G-protein-coupled receptor kinase 2 (GRK-2), which phosphorylates activated receptors, uncouples them from G
G protein-coupled receptor kinase 2 (GRK2) promotes breast tumorigenesis through a HDAC6-Pin1 axis
Nogues LR, et al.
EBioMedicine, 13, 132-145 (2016)
G protein-coupled receptor kinases: Crucial regulators of blood pressure
Yang J, et al.
Journal of the American Heart Association, 5(7), e003519-e003519 (2016)
G-protein-coupled receptor kinase 2 (GRK2) inhibitors: current trends and future perspectives
Guccione M, et al.
Journal of Medicinal Chemistry, 59(20), 9277-9294 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service