Skip to Content
Merck
All Photos(2)

Key Documents

SRP5024

Sigma-Aldrich

EIF2AK3 (563-1115), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DKFZp781H1925, HRI, PEK, PERK, WRS

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$641.00

$641.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$641.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

$641.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in E. coli

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

15-21 nmol/min·mg

mol wt

~115 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... EIF2AK3(9451)

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julie A Moreno et al.
Science translational medicine, 5(206), 206ra138-206ra138 (2013-10-11)
During prion disease, an increase in misfolded prion protein (PrP) generated by prion replication leads to sustained overactivation of the branch of the unfolded protein response (UPR) that controls the initiation of protein synthesis. This results in persistent repression of
Josef Neu
Gastroenterology and Nutrition null
Carsten Carlberg
Nutrigenomics null
H P Harding et al.
Molecular cell, 5(5), 897-904 (2000-07-06)
Malfolded proteins in the endoplasmic reticulum (ER) inhibit translation initiation. This response is believed to be mediated by increased phosphorylation of eukaryotic initiation factor 2alpha (eIF2alpha) and is hypothesized to reduce the work load imposed on the folding machinery during
Identification of common variants influencing risk of the tauopathy Progressive Supranuclear Palsy
Gunter U
Nature Genetics (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service