Skip to Content
Merck
All Photos(2)

Key Documents

SRP5002

Sigma-Aldrich

ALK3 (187-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

10q23del, ACVRLK3, CD292, SKR5

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$481.00

$481.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$481.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$481.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

1.0-1.4 nmol/min·mg

mol wt

~66 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... BMPR1A(657)

General description

BMPR1A (also known as bone morphogenetic protein receptor 1A) is a member of the transmembrane serine/threonine kinase family that include the type I receptors BMPR1A and BMPR1B and the type II receptor BMPR2. BMPR1A act as a minor susceptibility gene for PTEN-mutation-negative Cowden syndrome. BMPR1A regulates the PTEN protein levels by decreasing PTEN′s association with the degradative pathway. BMPR1A trafficking plays a significant role in FOP pathogenesis and is also involved in human T-cell differentiation.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristin A Waite et al.
Human molecular genetics, 12(6), 679-684 (2003-03-07)
The tumour suppressor gene PTEN encodes a dual-specificity phosphatase that recognizes protein and phosphatidylinositiol substrates and modulates cellular functions such as migration and proliferation. Germline mutations of PTEN have been shown to cause Cowden syndrome, Bannayan-Riley-Ruvalcaba syndrome and Proteus syndrome.
Teresa Cejalvo et al.
Immunology, 121(1), 94-104 (2007-04-12)
T-cell differentiation is driven by a complex network of signals mainly derived from the thymic epithelium. In this study we demonstrate in the human thymus that cortical epithelial cells produce bone morphogenetic protein 2 (BMP2) and BMP4 and that both

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service