Skip to Content
Merck
All Photos(1)

Key Documents

SRP4561

Sigma-Aldrich

Resistin from rat

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

ADSF, Adipose tissue-specific secretory factor, C/EBP-ε-regulated myeloid-specific secreted cysteine-rich protein, Cysteine-rich secreted protein A12-α-like 2, Cysteine-rich secreted protein FIZZ3, RSTN

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

rat

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

~11.9 kDa

packaging

pkg of 25 μg

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

Resistin is a new member to of the family of adipocyte-secreted proteins. The biological functions of resistin, as well as its molecular target, are largely unknown. Whether it promotes or antagonizes insulin action is still unclear. Recombinant rat Resistin is an 11.9 kDa protein consisting of 94 amino acid residues and 16 additional amino acids residues-His Tag.
Resistin is an adipocytokine secreted by fat tissues and is localized to pancreatic α-cells in rat.

Application

Resistin from rat has been used to study the effect of resistin on gonadotropins and local ovarian hormone-induced steroidogenesis.

Biochem/physiol Actions

Resistin mediates energy homeostasis and inflammation process. Upregulation of resistin is observed in obesity. It is also known to increase blood glucose level and might promote insulin resistance. Thus, it might influence type 2 diabetes. Resistin might be linked to insulin resistance mediated myocardial infarction (MI). In rats, overexpression of resistin is observed in myocardial infarction and cardiac dysfunction.

Physical form

Lyophilized from 20 mM Tris, pH 8.0.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 mg/mL. This solution can then be diluted into other aqueous buffers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maha M Sabry et al.
Archives of physiology and biochemistry, 126(5), 389-398 (2019-01-08)
Obesity and atherosclerosis are inflammatory states involving variable metabolic signals. The adipokine resistin is implicated in adipose tissue dysfunction and is modulated by PPARγ. In this study, resistin and PPARγ role is investigated in the development of CVS disease. Forty-eight
Regulatory role of gonadotropins and local factors produced by ovarian follicles on in vitro resistin expression and action on porcine follicular steroidogenesis
Rak A, et al.
Biology of Reproduction, 92(6), 142-141 (2015)
Resistin is produced by rat pancreatic islets and regulates insulin and glucagon in vitro secretion
Sassek M, et al.
Islets, 8(6), 177-185 (2016)
Sitagliptin decreases ventricular arrhythmias by attenuated glucose-dependent insulinotropic polypeptide (GIP)-dependent resistin signaling in infarcted rats
Lee TM, et al.
Bioscience Reports, BSR20150139-BSR20150139 (2016)
JingJing Shang et al.
iScience, 26(7), 107025-107025 (2023-06-30)
Osteogenic differentiation (OD) of bone marrow mesenchymal stem cells (BMSCs) contributes significantly to the regeneration of bone defects. Resistin, an adipose tissue-specific secretory factor, has been shown to involve many different functions, including metabolism, inflammation, cancer, and bone remodeling. However

Articles

Lipid Induced Insulin Resistance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service