Skip to Content
Merck
All Photos(1)

Key Documents

SRP4248

Sigma-Aldrich

MIP-1beta from mouse

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

CCL4, MIP-1- β, Macrophage inflammatory protein 1-β, Small inducible cytokine A4, T-cell activation protein 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

mouse

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

~7.8 kDa

packaging

pkg of 10 μg

impurities

endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Ccl4(20303)

General description

Mouse macrophage inflammatory protein-1β (MIP-1β) is a 7.8 kDa protein containing 69 amino acid residues. It plays an important role in the inflammatory response of blood monocytes and tissue macrophages.
MIP-1β is mainly generated by monocytes and lymphocytes. It is a member of CC family of chemokine intercrine cytokine. MIP-1β shares 70% similarity with MIP-1α. It is localized to a number of tissues such as smooth muscles, monocytes, neutrophils, mast cells, fibroblast B and T lymphocytes.[1]

Biochem/physiol Actions

Macrophage inflammatory protein-1 β (MIP-1β) promotes myelopoietic cellular growth and induces leukocyte chemoattraction. MIP-1β is an effective HIV infection suppressor.[1]

Physical form

Lyophilized without any additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 mg/mL. This solution can then be diluted into other aqueous buffers.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atlas of immunology, 318-318 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service