Skip to Content
Merck
All Photos(1)

Key Documents

SRP3323

Sigma-Aldrich

R-SPONDIN-3 human

recombinant, expressed in CHO cells, ≥95% (SDS-PAGE), ≥95% (HPLC)

Synonym(s):

PWTSR, RSPO3, Roof plate-specific spondin-3

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$450.00

$450.00


Check Cart for Availability


Select a Size

Change View
20 μG
$450.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$450.00


Check Cart for Availability

biological source

human

recombinant

expressed in CHO cells

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

mol wt

26.9 kDa

packaging

pkg of 20 μg

impurities

endotoxin, tested

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... RSPO3(84870)

General description

The R-Spondin (Rspo) proteins belong to the Rspo family of Wnt modulators. Currently, the family consists of four structurally related secreted ligands (Rspo 1-4), all containing furin-like and thrombospondin structural domains. The Rspo proteins can interact with the Frizzled/LRP6 receptor complex in a manner that causes the stabilization and resulting accumulation of the intracellular signaling protein, ?-catenin. This activity effectively activates and increases the subsequent nuclear signaling of ?-catenin. R-Spondin can also bind to the previously discovered G-protein coupled receptors, LGR-4 and LGR-5. Rspo/?-catenin signaling can act as an inducer of the transformed phenotype, and can also regulate the proliferation and differentiation of certain stem cell populations. Recombinant human R-Spondin-3 is a 26.9 kDa protein consisting of 240 amino acid residues. Due to glycosylation, R-Spondin-3 migrates at an apparent molecular weight of approximately 37.0 kDa by SDS PAGE analysis under reducing conditions.

Biochem/physiol Actions

The R-Spondin (Rspo) proteins belong to the Rspo family of Wnt modulators. Currently, the family consists of four structurally related secreted ligands (Rspo 1-4), all containing furin-like and thrombospondin structural domains. Recombinant human R-Spondin-3 is a 26.9 kDa protein consisting of 240 amino acid residues.

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 7.5 + 150 mM NaCl.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minke E Binnerts et al.
Proceedings of the National Academy of Sciences of the United States of America, 104(37), 14700-14705 (2007-09-07)
The R-Spondin (RSpo) family of secreted proteins act as potent activators of the Wnt/beta-catenin signaling pathway. We have previously shown that RSpo proteins can induce proliferative effects on the gastrointestinal epithelium in mice. Here we provide a mechanism whereby RSpo1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service