Skip to Content
Merck
All Photos(1)

Key Documents

SRP3306

Sigma-Aldrich

SCD40 LIGAND

Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

CD154, TNFSF5, TRAP, gp39

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$560.00

$560.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
$560.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

$560.00


Check Cart for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

5-10 ng/mL ED50

mol wt

16.3 kDa

packaging

pkg of 50 μg

impurities

endotoxin, tested

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TNFSF5(959)

General description

CD40, a member of the TNF receptor superfamily, is a cell surface protein expressed on B cells, dendritic cells, monocytes, thymic epithelial cells and, at low levels, on T cells. Signaling though CD40 plays an important role in the proliferation and differentiation of B cells, and is critical for immunoglobulin (Ig) class switching. The membrane-anchored CD40-Ligand is expressed almost exclusively on activated CD4+ T lymphocytes. Failure to express CD40L leads to “immunodeficiency with hyper-IgM”, a disease characterized by failure to produce IgG, IgA and IgE. The human CD40L gene codes for a 261 amino acid type II transmembrane protein, which contains a 22 amino acid cytoplasmic domain, a 24 amino acid transmembrane domain, and a 215 amino acid extracellular domain. The soluble form of CD40L is an 18 kDa protein comprising the entire TNF homologous region of CD40L and is generated in vivo by an intracellular proteolytic processing of the full length CD40L. Recombinant human soluble CD40 ligand is a 16.3 kDa protein containing 149 amino acid residues comprising the receptor binding TNF-like domain of CD40L.

Biochem/physiol Actions

CD40, a member of the TNF receptor superfamily, is a cell surface protein expressed on B cells, dendritic cells, monocytes, thymic epithelial cells and, at low levels, on T cells. The soluble form of CD40L is an 18 kDa protein comprising the entire TNF homologous region of CD40L and is generated in vivo by an intracellular proteolytic processing of the full length CD40L. Recombinant human soluble CD40 ligand is a 16.3 kDa protein containing 149 amino acid residues comprising the receptor binding TNF-like domain of CD40L.

Physical form

Lyophilized from 10mM Sodium Phosphate, pH 7.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Contribution of surface localization to platelet activation by CD154 antibodies and introduction of a novel approach for studying immune complex activity.
Meghan Hatfield et al.
Thrombosis research, 127(6), 571-575 (2010-01-20)
D Hollenbaugh et al.
The EMBO journal, 11(12), 4313-4321 (1992-12-01)
Signals delivered to B cells via CD40 can synergize with those provided by other B cell surface receptors to induce B cell proliferation and antibody class switching as well as modulate cytokine production and cell adhesion. Recently, it has been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service