Skip to Content
Merck
All Photos(1)

Key Documents

SRP3289

Sigma-Aldrich

IL-36 GAMMA human

≥95% (SDS-PAGE & HPLC), recombinant, expressed in E. coli, lyophilized

Synonym(s):

IL-1ε, IL-1F, IL-1H1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$459.00

$459.00


Check Cart for Availability


Select a Size

Change View
10 μG
$459.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$459.00


Check Cart for Availability

Product Name

IL-36 GAMMA human, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

mol wt

17.0 kDa

packaging

pkg of 10 μg

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL36B(27177)

General description

IL-36γ is highly expressed in psoriatic plaques and in tissues containing epithelial cells. IL-36γ signals through the IL-1Rrp2 (IL-1R6) receptor, which is primarily expressed on certain dendritic cells. The interaction of the IL-1Rrp2 receptor with IL-36 ligands induces dendritic cell maturation and activation. IL-36γ also functions as an agonist of NF-kappaB, and can stimulate the inflammatory response in bronchial epithelial cells.

Biochem/physiol Actions

The IL-1 family is comprised of 11 structurally related ligands, including recently re-named IL-36 α (IL-1F6), β (IL-1F8) and γ (IL-1F9). Recombinant human IL-36γ is a 17.0 kDa protein containing 152 amino acid residues.

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 7.0 + 50 mM NaCl.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dinarello, C. et al.
Nature Immunol., 11, 973-973 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service