Skip to Content
Merck
All Photos(1)

Key Documents

SRP3244

Sigma-Aldrich

IL-7 from rat

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

Lymphopoietin 1(LP-1), pre-B cell factor

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$442.00

$442.00


Estimated to ship on26 May 2025



Select a Size

Change View
10 μG
$442.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$442.00


Estimated to ship on26 May 2025


biological source

rat

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

≤0.2 ng/mL ED50

mol wt

15.0 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... IL7(25647)

General description

IL-7 is a hematopoietic growth factor, which affects primarily early B and T cells. Produced by thymic stromal cells, spleen cells and keratinocytes, IL-7 can also co-stimulate the proliferation of mature T cells in combination with other factors such as ConA and IL-2. Human and murine IL-7 is cross-species reactive. Recombinant rat IL-7 is a 15.0 kDa protein containing 130 amino acid residues.

Biochem/physiol Actions

IL-7 is a hematopoietic growth factor, which affects primarily early B and T cells. Recombinant rat IL-7 is a 15.0 kDa protein containing 130 amino acid residues.

Sequence

MDCHIKDKDG KAFGSVLMIS INQLDKMTGT DSDCPNNEPN FFKKHLCDDT KEAAFLNRAA RKLRQFLKMN ISEEFNDHLL RVSDGTQTLV NCTSKEEKTI KEQKKNDPCF LKRLLREIKT CWNKILKGSI

Physical form

Lyophilized from 10 mM Sodium Acetate, pH 5.8.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spits, H.
Cytokine Reference, 137-153 (2001)
A E Namen et al.
The Journal of experimental medicine, 167(3), 988-1002 (1988-03-01)
We have used a biological assay system we developed to biochemically purify a previously uncharacterized murine lymphopoietic growth factor designated lymphopoietin 1 (LP-1). This factor is capable of stimulating the proliferation and extended maintenance of precursor cells of the B
Nicholas J Steers et al.
Vaccine, 32(28), 3509-3516 (2014-05-06)
The flanking amino acids that surround epitopes are critical for effective antigen processing and maintenance of epitope integrity. In the present study, the frequency and characteristics of each amino acid that flanked the peptides generated from the proteasomal degradation of
Antoine Chéret et al.
The Journal of antimicrobial chemotherapy, 70(7), 2108-2120 (2015-04-23)
Therapeutic control of HIV replication reduces the size of the viral reservoir, particularly among central memory CD4+ T cells, and this effect might be accentuated by early treatment. We examined the effect of ART initiated at the time of the
Sabine Schmied et al.
Immunology, 145(4), 558-569 (2015-04-18)
The Wilms' tumour-1 (WT1) protein is considered a prime target for cancer immunotherapy based on its presumptive immunogenicity and widespread expression across a variety of malignancies. However, little is known about the naturally occurring WT1-specific T-cell repertoire because self-derived antigens

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service