Skip to Content
Merck
All Photos(1)

Key Documents

SRP3243

Sigma-Aldrich

IL-3 beta from rat

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

HCGF, MCGF (Mast cell growth factor), Multi-CSF, P-cell stimulation factor

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$442.00

$442.00


Estimated to ship on27 April 2025



Select a Size

Change View
20 μG
$442.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$442.00


Estimated to ship on27 April 2025


biological source

rat

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

≤10 ng/mL ED50

mol wt

16.3 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... IL3(24495)

General description

IL-3 is a hematopoietic growth factor that promotes the survival, differentiation and proliferation of committed progenitor cells of the megakaryocyte, granulocyte-macrophage, erythroid, eosinophil, basophil and mast cell lineages. Produced by T cells, mast cells and eosinophils, IL-3 enhances thrombopoieses, phagocytosis, and antibody-mediated cellular cytotoxicity. Its ability to activate monocytes suggests that IL-3 may have additional immunoregulatory roles. Many of the IL-3 activities depend upon co-stimulation with other cytokines. IL-3 is species-specific, variably glycosylated cytokine. Recombinant rat IL-3β is a 16.3 kDa globular protein containing 144 amino acid residues.

Biochem/physiol Actions

IL-3 is a hematopoietic growth factor that promotes the survival, differentiation and proliferation of committed progenitor cells of the megakaryocyte, granulocyte-macrophage, erythroid, eosinophil, basophil and mast cell lineages. Recombinant rat IL-3β is a 16.3 kDa globular protein containing 144 amino acid residues.

Sequence

MISDRGSDAH HLLRTLDCRT IALEILVKLP YPQVSGLNNS DDKANLRNST LRRVNLDEFL KSQEEFDSQD TTDIKSKLQK LKCCIPAAAS DSVLPGVYNK DLDDFKKKLR FYVIHLKDLQ PVSVSRPPQP TSSSDNFRPM TVEC

Physical form

Lyophilized from 5mM Sodium Citrate, pH 4.0.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In Peptide Growth Factors and their Receptors I.
Ihle, J.N.
Peptide Growth Factors and their Receptors I, 541-575 (1991)
J N Ihle
Chemical immunology, 51, 65-106 (1992-01-01)
Considerable information has accumulated over the past several years relating to the biological activities of IL-3 which supports that concept that IL-3 has a unique capacity to support the proliferation and differentiation of a wide spectrum of hematopoietic cells. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service