Skip to Content
Merck
All Photos(1)

Key Documents

SRP3236

Sigma-Aldrich

TPO from mouse

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

MGDF, Megakaryocyte colony-stimulating factor, Thrombopoietin, c-MPL Ligand

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$442.00

$442.00


Estimated to ship on02 April 2025



Select a Size

Change View
10 μG
$442.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$442.00


Estimated to ship on02 April 2025


biological source

mouse

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

<1.0 ng/mL ED50

mol wt

18.7 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... THPO(21832)

General description

TPO (thrombopoietin) is a hematopoietic cytokine expressed in multiple tissue types. It is predominantly produced by liver and kidney and acts as a ligand for the cell surface receptor c-mpl.[1][2]
TPO is a lineage specific growth factor, produced in the liver, kidney and skeletal muscle. It stimulates the proliferation and maturation of megakaryocytes, and promotes increased circulating levels of platelets in vivo. TPO signals through the c-mpl receptor and acts as an important regulator of circulating platelets. Human and murine TPO exhibits cross-species reactivity. Recombinant murine TPO is a fully biologically active 174 amino acid polypeptide (18.7 kDa), which contains the erythropoietin-like domain of the full length TPO protein.

Biochem/physiol Actions

TPO (thrombopoietin) is the key regulator of thrombopoiesis for platelet lineage. In both mice and humans, inactivation of this gene results in significant decrease in number of mature platelets produced by marrow progenitors.[1] Transgenic mice with increased expression of this protein show progressive fibrosis and osteosclerosis of the bone marrow, as well as histological changes in kidneys, leading to mesangioproliferative glomerulopathy.[2]

Sequence

SPVAPACDPR LLNKLLRDSH LLHSRLSQCP DVDPLSIPVL LPAVDFSLGE WKTQTEQSKA QDILGAVSLL LEGVMAARGQ LEPSCLSSLL GQLSGQVRLL LGALQGLLGT QLPLQGRTTA HKDPNALFLS LQQLLRGKVR FLLLVEGPTL CVRRTLPTTA VPSSTSQLLT LNKF

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 8.0.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of ?0.5 mg/ml. Slow to dissolve. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bryan McIntosh et al.
Experimental hematology, 36(7), 799-806 (2008-04-16)
Platelet production is regulated primarily by the cytokine thrombopoietin (TPO). Although TPO is expressed in several different tissues, only in the bone marrow has the level of expression been reported to increase in response to reduced numbers of platelets. In
Haruko K Shimoda et al.
American journal of hematology, 82(9), 802-806 (2007-06-05)
We previously reported that mice transgenic (Tg) for thrombopoietin (TPO) developed progressive fibrosis and osteosclerosis of the bone marrow. Here, we show that TPO-overexpressing mice also exhibited notable histological changes in the kidneys, including an increased number of mesangial cells
Kuter, D.J., et al.
Cytokine Reference, 965-982 (2001)
Krasimira Rozenova et al.
Blood, 125(11), 1730-1738 (2015-02-01)
Hematopoietic stem cell (HSC) self-renewal and multilineage reconstitution are controlled by positive and negative signaling cues with perturbations leading to disease. Lnk is an essential signaling adaptor protein that dampens signaling by the cytokine thrombopoietin (Tpo) to limit HSC expansion.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service