Skip to Content
Merck
All Photos(1)

Key Documents

SRP3198

Sigma-Aldrich

flt3-Ligand from mouse

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

Fms-related tyrosine kinase 3 ligand

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$500.00

$500.00


Available to ship on09 April 2025Details


Request a Bulk Order

Select a Size

Change View
10 μG
$500.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$500.00


Available to ship on09 April 2025Details


Request a Bulk Order

biological source

mouse

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

5.0-8.0 ng/mL

mol wt

18.6 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... FLT3LG(14256)

Related Categories

General description

FMS-like tyrosine kinase 3 ligand (Flt3l), a cytokine, is also termed as Flk2/CD135.[1] It is a hematopoietic growth factor.[2] Recombinant murine Flt3-ligand is a soluble 18.6 kDa protein consisting of 163 amino acid residues.

Biochem/physiol Actions

Flt3l (FMS-like tyrosine kinase 3 ligand) acts as a stimulant of hematopoietic progenitor cell proliferation. It facilitates the commitment of lymphoid lineage, and aids in dendritic cell (DC) development. It is involved in myoblast differentiation, and drives skeletal muscle regeneration. It acts on p120RasGAP (rat sarcoma GTPase activating protein) and suppresses Erk (extracellular signal-regulated kinase) signaling, which in turn promotes myogenesis.[2] In lymphoid tissues, it modulates DC homeostasis, which is facilitated by steady state as well as proliferative T-cells.[3] Recombinant murine Flt3-Ligand is a soluble 18.6 kDa protein consisting of 163 amino acid residues.
Fms-like tyrosine kinase 3 (FLT3) ligand (FL), expressed by leukemic cells may help to increase the multiplication through an autocrine process.[4]

Sequence

MTPDCYFSHS PISSNFKVKF RELTDHLLKD YPVTVAVNLQ DEKHCKALWS LFLAQRWIEQ LKTVAGSKMQ TLLEDVNTEI HFVTSCTFQP LPECLRFVQT NISHLLKDTC TQLLALKPCI GKACQNFSRC LEVQCQPDSS TLLPPRSPIA LEATELPEPR PRQ

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 7.8.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

FLT3 ligand plasma levels in acute myeloid leukemia.
Pierre Peterlin et al.
Haematologica, 104(6), e240-e243 (2019-01-12)
Panagiotis Tsapogas et al.
International journal of molecular sciences, 18(6) (2017-05-26)
The cytokine Fms-like tyrosine kinase 3 ligand (FL) is an important regulator of hematopoiesis. Its receptor, Flt3, is expressed on myeloid, lymphoid and dendritic cell progenitors and is considered an important growth and differentiation factor for several hematopoietic lineages. Activating
Yasuyuki Saito et al.
European journal of immunology, 43(6), 1651-1658 (2013-03-23)
Lymphoid-tissue dendritic cells (DCs) are short-lived and need to be continuously replenished from bone marrow-derived DC progenitor cells. Fms-related tyrosine kinase 3 is expressed during cellular development from hematopoietic progenitors to lymphoid-tissue DCs. Fms-related tyrosine kinase 3 ligand (Flt3L) is
Yejing Ge et al.
Journal of cell science, 126(Pt 15), 3370-3379 (2013-05-25)
Various cues initiate multiple signaling pathways to regulate the highly coordinated process of skeletal myogenesis. Myoblast differentiation comprises a series of ordered events starting with cell cycle withdrawal and ending with myocyte fusion, with each step probably controlled by multiple
K Mackarehtschian et al.
Immunity, 3(1), 147-161 (1995-07-01)
The flk2 receptor tyrosine kinase has been implicated in hematopoietic development. Mice deficient in flk2 were generated. Mutants developed into healthy adults with normal mature hematopoietic populations. However, they possessed specific deficiencies in primitive B lymphoid progenitors. Bone marrow transplantation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service