Skip to Content
Merck
All Photos(1)

Key Documents

SRP3175

Sigma-Aldrich

TL-1A human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

TNFSF15, VEGI

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$448.00

$448.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
20 μG
$448.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$448.00


Check Cart for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

10.0-100.0 ng/mL

mol wt

22.0 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TNFSF15(9966)

General description

TL-1A belongs to the TNF superfamily of ligands. It is expressed predominantly in endothelial cells, and to a lesser extent in placenta, lung, kidney, skeletal muscle, pancreas, small intestine and colon. TL-1A inhibits endothelial cell proliferation and angiogenesis, and has been shown to induce NFkB activation, caspase activity, and apoptosis in responding cell lines. TL-1A interacts with TNFRSF25/DR3 receptor, but can also bind to a decoy receptor TNFRSF21/DR6. Recombinant human TL-1A is a 22.0 kDa polypeptide of 194 amino acid residues.

Biochem/physiol Actions

TL-1A belongs to the TNF superfamily of ligands. Recombinant human TL-1A is a 22.0 kDa polypeptide of 194 amino acid residues.

Sequence

QLRAQGEASV QFQALKGQEF APSHQQVYAP LRADGDKPRA HLTVVRQTPT QHFKNQFPAL HWEHELGLAF TKNRMNYTNK FLLIPESGDY FIYSQVTFRG MTSECSEIRQ AGRPNKPDSI TVVITKVTDS YPEPTQLLMG TKSVCEVGSN WFQPIYLGAM FSLQEGDKLM VNVSDISLVD YTKEDKTFFG AFLL

Physical form

Lyophilized from 10 mM Tris, pH 8.5 + 75 mM Arginine.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoon-Joong Kang et al.
Cytokine, 29(5), 229-235 (2005-03-12)
TL1A (VEGI/TNFSF15) is the ligand for DR3 (TNFRSF12) and is a newly identified member of the tumor necrosis factor superfamily (TNFSF). Previously, DR3 has been shown to have a role in atherogenesis through stimulation of matrix degrading enzymes including matrix
Li-Jin Chew et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 16(7), 742-744 (2002-03-30)
Vascular endothelial cell growth inhibitor (VEGI), a member of the tumor necrosis factor (TNF) family, is an endothelial cell-specific inhibitor of angiogenesis. Overexpression by cancer cells of a secretable VEGI fusion protein resulted in abrogation of xenograft tumor progression, but

Questions

  1. Hello, Is this monomer or Trimer version? Also could you give me the sequence? Than you

    1 answer
    1. This material has not been tested to determine the structure. The sequence is available is the description section of the product page.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service