Skip to Content
Merck
All Photos(1)

Key Documents

SRP3168

Sigma-Aldrich

TECK (CCL25) human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

CCL25, Thymus Expressed Chemokine

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$457.00

$457.00


Check Cart for Availability


Select a Size

Change View
20 μG
$457.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$457.00


Check Cart for Availability

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

1.0-10.0 ng/mL

mol wt

14.2 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCL25(6370)

General description

TECK is a CC chemokine, specifically expressed by thymic stromal cells, and signals through the CCR9 receptor. TECK is chemotactic towards activated macrophages, thymocytes and dendritic cells. Recombinant human TECK is a 14.2 kDa protein containing 127 amino acid residues, including the four conserved cysteine residues present in CC chemokines.

Biochem/physiol Actions

TECK is a CC chemokine, specifically expressed by thymic stromal cells, and signals through the CCR9 receptor. Recombinant human TECK is a 14.2 kDa protein containing 127 amino acid residues, including the four conserved cysteine residues present in CC chemokines.

Sequence

QGVFEDCCLA YHYPIGWAVL RRAWTYRIQE VSGSCNLPAA IFYLPKRHRK VCGNPKSREV QRAMKLLDAR NKVFAKLHHN MQTFQAGPHA VKKLSSGNSK LSSSKFSNPI SSSRKNVSLL ISANSGL

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Zaballos et al.
Journal of immunology (Baltimore, Md. : 1950), 162(10), 5671-5675 (1999-05-07)
Thymus-expressed chemokine (TECK) has been reported to chemoattract dendritic cells, thymocytes, and activated macrophages. Here, we show that TECK is a specific agonist for a human orphan receptor called GPR-9-6. We have determined the cDNA sequence of human GPR-9-6 and
A P Vicari et al.
Immunity, 7(2), 291-301 (1997-08-01)
A novel CC chemokine was identified in the thymus of mouse and human and was designated TECK (thymus-expressed chemokine). TECK has weak homology to other CC chemokines and maps to mouse chromosome 8. Besides the thymus, mRNA encoding TECK was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service