Skip to Content
Merck
All Photos(1)

Key Documents

SRP3165

Sigma-Aldrich

sTRAIL Receptor-2 (DR5) human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

DR5, KILER, TNFRSF10B, soluble TRAIL Receptor-2

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$381.00

$381.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
$381.00

About This Item

UNSPSC Code:
51111800
NACRES:
NA.32

$381.00


Check Cart for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

1000 ng/mL

mol wt

14.9 kDa

packaging

pkg of 50 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

DR5 (death receptor 5) or TNFRSF10B (tumor necrosis factor receptor superfamily member 10b) is a member of the tumor necrosis factor receptor (TNFR) superfamily.[1] It is a tumor suppressor gene localized to human chromosome 8p and is composed of 10 coding exons. The encoded protein is an integral plasma membrane protein.[2]
Recombinant human sTNFRII is an 18.9 kDa protein (174 amino acid residues) comprising the cysteine rich ligand binding portion of the extracellular domain of the TNFRII protein.

Biochem/physiol Actions

It initiates the formation of death-inducing signal complex (DISC) by recruiting fas-associated death domain (FADD) and caspases to its cytoplasmic death domain (DD). In human lung cancer cells, parthenolide (PTL) induces extrinsic apoptosis by up-regulating TNFRSF10B.[1] Deregulation of this gene results in metastasis, invasion and migration of tumor cells. Mutation leads to loss of p arm in this gene which is frequently the case in head and neck tumors.[2] In a breast cancer xenograft mouse model, DR5 facilitates skeletal metastasis.[3] In prostate carcinoma (PCa) and prostatic intraepithelial neoplasia (PIN), the expression of DR5 is lower in higher grade tumors.[4]

Sequence

MESALITQQD LAPQQRVAPQ QKRSSPSEGL CPPGHHISED GRDCISCKYG QDYSTHWNDL LFCLRCTRCD SGEVELSPCT TTRNTVCQCE EGTFREEDSP EMCRKCRTGC PRGMVKVGDC TPWSDIECVH KES

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Death receptor 5 expression is inversely correlated with prostate cancer progression.
Molecular Medicine Reports, 10(5), 2279-2286 (2014)
TRAIL-R2 promotes skeletal metastasis in a breast cancer xenograft mouse model.
Oncotarget, 6(11), 9502-9516 (2015)
Parthenolide induces apoptosis via TNFRSF10B and PMAIP1 pathways in human lung cancer cells.
Zhao X et al
Journal of Experimental & Clinical Cancer Research, 33, 3-3 (2014)
Tumor necrosis factor receptor superfamily 10B (TNFRSF10B): an insight from structure modeling to virtual screening for designing drug against head and neck cancer.
Tahir RA et al
Theoretical Biology & Medical Modelling, 10(38), 1742-4682 (2013)
Ashkenazi, A., et al.
Cytokine Reference, 1733-1742 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service