Skip to Content
Merck
All Photos(1)

Key Documents

SRP2127

Sigma-Aldrich

TR, α1, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE)

Synonym(s):

AR7, EAR7, ERB-T-1, ERBA, ERBA1, MGC000261, MGC43240, NR1A1, THRA1, c-ERBA-1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$370.00

$370.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
$370.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

$370.00


Check Cart for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

frozen liquid

mol wt

~81.5 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

350 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... THRA(7067)

Biochem/physiol Actions

Nuclear receptors form the largest known family of transcription factors and have a crucial role in nearly all aspects of vertebrate development and adult physiology by transducing the effects of hormones into transcriptional responses. The family is defined by two domains: (a) the central, highly conserved, DNA-binding domain (DBD) of approx. 66 amino acids, and (b) the C-terminal, structurally conserved, ligand-binding domain (LBD) of approximately 250 amino acids. The amino-terminal regions are least conserved among nuclear receptor sequences. This domain is highly divergent between the TR and TR isoforms, which suggests differential roles in transcriptional regulation. In addition, alternative splicing of the TR gene generates two isoforms, TR 1 and TR 2 with completely different amino-terminal domains. Unliganded TR inhibits the formation of a functional pre-initiation complex through direct interaction with TBP and transcription factor IIB. Additionally, in the absence of ligand, TR has been shown to repress transcription through recruitment of a corepressor complex, which also includes Sin3A and histone deacetylase. Ligand binding releases the corepressor complex and recruits a coactivator complex that includes multiple histone acetyltransferases, including a steroid receptor family coactivator, p300/CREB-binding protein-associated factor (PCAF), and CREB binding protein (CBP).

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Differential recognition of target genes by nuclear receptor monomers, dimers, and heterodimers.
C K Glass
Endocrine reviews, 15(3), 391-407 (1994-06-01)
D Moras et al.
Current opinion in cell biology, 10(3), 384-391 (1998-06-26)
In the past few years our understanding of nuclear receptor action has dramatically improved as a result of the elucidation of the crystal structures of the empty (apo) ligand-binding domains of the nuclear receptor and of complexes formed by the
The nuclear receptor superfamily: the second decade.
D J Mangelsdorf et al.
Cell, 83(6), 835-839 (1995-12-15)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service